Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133727.2 | NP_598488.2 | 7 - 8 | 786 | 112 | |
GenBank mRNA | AK009746.1 | - | 7 - 8 | 767 | 112 | |
BC018316.1 | - | 7 - 8 | 754 | 4160465 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |