Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133789.3 | NP_598550.2 | 8 - 9 | 1618 | 120 | |
GenBank mRNA | AF414080.1 | - | 8 - 9 | 1618 | 120 | |
BC080283.1 | - | 8 - 9 | 1184 | 6401782 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |