Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700021419.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021419.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021419.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021423.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021423.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021423.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021424.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021424.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021424.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021426.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021426.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021426.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021418.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021418.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021418.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021422.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021422.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021422.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021427.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021427.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021427.mm.1 |
NM_172739.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021420.mm.1 |
TC0700002430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021420.mm.1 |
NM_172739.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021420.mm.1 |
NM_172739.4 |
Refseq |