Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027725.3 | NP_082001.2 | 8 - 9 | 777 | 64 | |
NR_027636.1 | 11 - 12 | 1072 | 64 | |||
GenBank mRNA | AK016201.1 | - | 11 - 12 | 1067 | 64 | |
AK016977.1 | - | 8 - 9 | 778 | 64 | ||
BC049704.1 | - | 8 - 9 | 777 | 64 | ||
BC144994.1 | - | 8 - 9 | 754 | 64 | ||
BC145995.1 | - | 8 - 9 | 777 | 40131298 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |