Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011978.2 | NP_036108.2 | 7 - 8 | 1584 | 61 | |
GenBank mRNA | AF033031.1 | - | 7 - 8 | 1465 | 61 | |
AF072757.1 | - | 7 - 8 | 1462 | 61 | ||
AJ223958.1 | - | 7 - 8 | 1502 | 61 | ||
AK144069.1 | - | 7 - 8 | 1586 | 61 | ||
AK144070.1 | - | 7 - 8 | 1498 | 61 | ||
BC013442.1 | - | 7 - 8 | 1567 | 4236616 | 61 | |
BC022170.1 | - | 7 - 8 | 1563 | 4159081 | 61 | |
BC024735.1 | - | 7 - 8 | 1553 | 5123650 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |