Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252543.1 | NP_001239472.1 | 16 - 17 | 2022 | 105 | |
NM_001252544.1 | NP_001239473.1 | 16 - 17 | 2015 | 105 | ||
NM_001252545.1 | NP_001239474.1 | 16 - 17 | 2004 | 105 | ||
NM_001281816.1 | NP_001268745.1 | 13 - 14 | 1644 | 105 | ||
NM_019787.4 | NP_062761.2 | 16 - 17 | 2033 | 105 | ||
GenBank mRNA | AF200326.1 | - | 15 - 16 | 1905 | 105 | |
AK014597.1 | - | 16 - 17 | 1983 | 105 | ||
AK029661.1 | - | 13 - 14 | 1645 | 105 | ||
AK139729.1 | - | 16 - 17 | 1948 | 105 | ||
AK146083.1 | - | 16 - 17 | 1982 | 105 | ||
AK149693.1 | - | 16 - 17 | 1962 | 105 | ||
AK153214.1 | - | 16 - 17 | 1989 | 105 | ||
AK166330.1 | - | 16 - 17 | 1965 | 105 | ||
AK167382.1 | - | 16 - 17 | 1970 | 105 | ||
AK167732.1 | - | 16 - 17 | 1951 | 105 | ||
AK171597.1 | - | 16 - 17 | 1963 | 105 | ||
AK199769.1 | - | 2 - 3 | 203 | 105 | ||
BC005464.1 | - | 16 - 17 | 1958 | 3591420 | 105 | |
BC011160.1 | - | 16 - 17 | 1985 | 4024353 | 105 | |
CK623414.1 | - | 2 - 3 | 254 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |