Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252543.1 | NP_001239472.1 | 7 - 8 | 946 | 66 | |
NM_001252544.1 | NP_001239473.1 | 7 - 8 | 939 | 66 | ||
NM_001252545.1 | NP_001239474.1 | 7 - 8 | 928 | 66 | ||
NM_001281816.1 | NP_001268745.1 | 4 - 5 | 568 | 66 | ||
NM_019787.4 | NP_062761.2 | 7 - 8 | 957 | 66 | ||
GenBank mRNA | AF200326.1 | - | 6 - 7 | 829 | 66 | |
AK014597.1 | - | 7 - 8 | 907 | 66 | ||
AK029661.1 | - | 4 - 5 | 569 | 66 | ||
AK139729.1 | - | 7 - 8 | 872 | 66 | ||
AK146083.1 | - | 7 - 8 | 906 | 66 | ||
AK149693.1 | - | 7 - 8 | 886 | 66 | ||
AK153214.1 | - | 7 - 8 | 913 | 66 | ||
AK158398.1 | - | 7 - 8 | 924 | 66 | ||
AK166330.1 | - | 7 - 8 | 889 | 66 | ||
AK167382.1 | - | 7 - 8 | 895 | 66 | ||
AK167732.1 | - | 7 - 8 | 875 | 66 | ||
AK171597.1 | - | 7 - 8 | 887 | 66 | ||
AK184652.1 | - | 1 - 2 | 40 | 66 | ||
BC005464.1 | - | 7 - 8 | 882 | 3591420 | 66 | |
BC011160.1 | - | 7 - 8 | 909 | 4024353 | 66 | |
BU614902.1 | - | 3 - 4 | 286 | 66 | ||
CF724836.1 | - | 1 - 2 | 136 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |