Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183208.3 | NP_899031.2 | 19 - 20 | 2978 | 137 | |
GenBank mRNA | BC057691.1 | - | 9 - 10 | 1215 | 137 | |
BC058646.1 | - | 19 - 20 | 2878 | 6405777 | 137 | |
BC065120.1 | - | 19 - 20 | 2871 | 6856060 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |