Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172673.3 | NP_766261.2 | 13 - 14 | 1356 | 66 | |
GenBank mRNA | AK030883.1 | - | 13 - 14 | 1112 | 66 | |
AK044270.1 | - | 13 - 14 | 1107 | 66 | ||
AK146383.1 | - | 12 - 13 | 1236 | 66 | ||
BC038623.1 | - | 13 - 14 | 1093 | 66 | ||
BC057549.1 | - | 10 - 11 | 885 | 6416914 | 66 | |
BC062889.1 | - | 13 - 14 | 1313 | 5698263 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |