Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205369.1 | NP_001192298.1 | 4 - 5 | 135 | ||
NM_001205370.1 | NP_001192299.1 | 4 - 5 | 135 | |||
NM_001205371.1 | NP_001192300.1 | 4 - 5 | 135 | |||
NM_177054.5 | NP_796028.2 | 4 - 5 | 135 | |||
NM_199038.3 | NP_950239.2 | 4 - 5 | 135 | |||
GenBank mRNA | AK131751.1 | - | 4 - 5 | 135 | ||
AK160708.1 | - | 4 - 5 | 135 | |||
AY484583.1 | - | 4 - 5 | 135 | |||
AY484585.1 | - | 4 - 5 | 135 | |||
BC034556.1 | - | 4 - 5 | 135 | |||
BC064446.1 | - | 4 - 5 | 6843743 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |