Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002894.2 | NP_001002894.2 | 1 - 2 | 78 | ||
GenBank mRNA | AY673647.1 | - | 1 - 2 | 78 | ||
BC141199.1 | - | 1 - 2 | 9055811 | 78 | ||
CJ053047.1 | - | 1 - 2 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |