Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009502.4 | NP_033528.3 | 4 - 5 | 590 | 91 | |
GenBank mRNA | AK077850.1 | - | 4 - 5 | 626 | 91 | |
AK135530.1 | - | 4 - 5 | 626 | 91 | ||
BC008520.1 | - | 4 - 5 | 590 | 3495710 | 91 | |
L18880.1 | - | 4 - 5 | 519 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |