Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205241.1 | NP_001192170.1 | 2 - 3 | 930 | 78 | |
NM_017479.3 | NP_059507.2 | 2 - 3 | 1054 | 78 | ||
GenBank mRNA | AF222800.1 | - | 2 - 3 | 970 | 78 | |
AK045188.1 | - | 2 - 3 | 1066 | 78 | ||
AK083123.1 | - | 2 - 3 | 1069 | 78 | ||
AK135465.1 | - | 2 - 3 | 1106 | 78 | ||
AK147509.1 | - | 2 - 3 | 931 | 78 | ||
BC095974.1 | - | 1 - 2 | 707 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |