Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198167.3 | NP_937810.2 | 16 - 17 | 1642 | 64 | |
GenBank mRNA | AK090200.1 | - | 16 - 17 | 1563 | 64 | |
AK159673.1 | - | 16 - 17 | 1647 | 64 | ||
BC039277.1 | - | 5 - 6 | 624 | 64 | ||
BC057136.1 | - | 5 - 6 | 605 | 6529222 | 64 | |
BC059283.1 | - | 9 - 10 | 990 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |