Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198167.3 | NP_937810.2 | 22 - 23 | 2348 | 72 | |
GenBank mRNA | AK090200.1 | - | 22 - 23 | 2269 | 72 | |
AK159673.1 | - | 22 - 23 | 2353 | 72 | ||
BC026370.1 | - | 5 - 6 | 607 | 72 | ||
BC039277.1 | - | 11 - 12 | 1330 | 72 | ||
BC057136.1 | - | 11 - 12 | 1311 | 6529222 | 72 | |
BC059283.1 | - | 15 - 16 | 1696 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |