Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162864.1 | NP_001156336.1 | 10 - 11 | 1321 | 60 | |
GenBank mRNA | AK144536.1 | - | 9 - 10 | 1055 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Inventoried Probe spans exons |