Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053250.2 | NP_444480.1 | 3 - 4 | 201 | 86 | |
NM_181664.2 | NP_858050.1 | 3 - 4 | 201 | 86 | ||
GenBank mRNA | AF367970.1 | - | 3 - 4 | 201 | 86 | |
AF367971.1 | - | 3 - 4 | 201 | 86 | ||
AK080272.1 | - | 3 - 4 | 201 | 86 | ||
AY555745.1 | - | 3 - 4 | 192 | 86 | ||
BC116360.1 | - | 3 - 4 | 201 | 40099973 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |