Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177231.2 | NP_796205.1 | 9 - 10 | 997 | 81 | |
NM_178220.3 | NP_835738.1 | 9 - 10 | 997 | 81 | ||
GenBank mRNA | AK052588.1 | - | 9 - 10 | 774 | 81 | |
AK090090.1 | - | 9 - 10 | 824 | 81 | ||
AK144054.1 | - | 9 - 10 | 796 | 81 | ||
AK147508.1 | - | 9 - 10 | 998 | 81 | ||
AK147566.1 | - | 9 - 10 | 1019 | 81 | ||
AK165514.1 | - | 9 - 10 | 796 | 81 | ||
BC108969.1 | - | 9 - 10 | 764 | 40047224 | 81 | |
BC108970.1 | - | 9 - 10 | 764 | 40047225 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |