Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700016949.mm.1 |
TC0700001939.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016949.mm.1 |
NM_029609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016949.mm.1 |
NM_029609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016944.mm.1 |
TC0700001939.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016944.mm.1 |
NM_029609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016944.mm.1 |
NM_029609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016945.mm.1 |
TC0700001939.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016945.mm.1 |
NM_029609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016945.mm.1 |
NM_029609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016950.mm.1 |
TC0700001939.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016950.mm.1 |
NM_029609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016950.mm.1 |
NM_029609.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016948.mm.1 |
TC0700001939.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700016948.mm.1 |
NM_029609.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700016948.mm.1 |
NM_029609.1 |
Refseq |