Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081038.2 | NP_001074507.2 | 13 - 14 | 1553 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |