Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081038.2 | NP_001074507.2 | 3 - 4 | 558 | 121 | |
GenBank mRNA | AK087912.1 | - | 3 - 4 | 560 | 121 | |
BB666983.1 | - | 3 - 4 | 559 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |