Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026391.2 | NP_080667.1 | 4 - 5 | 483 | 89 | |
GenBank mRNA | AF366393.1 | - | 4 - 5 | 391 | 89 | |
AK005033.1 | - | 3 - 4 | 372 | 89 | ||
AK011198.1 | - | 4 - 5 | 505 | 89 | ||
AK129386.2 | - | 4 - 5 | 510 | 89 | ||
BC066022.1 | - | 4 - 5 | 483 | 6828707 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |