Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026391.2 | NP_080667.1 | 6 - 7 | 779 | 93 | |
GenBank mRNA | AK005033.1 | - | 5 - 6 | 668 | 93 | |
AK011198.1 | - | 6 - 7 | 801 | 93 | ||
BC066022.1 | - | 6 - 7 | 779 | 6828707 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |