Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146119.1 | NP_001139591.1 | 10 - 11 | 1094 | 86 | |
NM_029758.4 | NP_084034.2 | 11 - 12 | 1167 | 86 | ||
GenBank mRNA | AK010821.1 | - | 9 - 10 | 1030 | 86 | |
AK047142.1 | - | 10 - 11 | 1057 | 86 | ||
BC049664.1 | - | 11 - 12 | 1129 | 6532813 | 86 | |
BC052465.1 | - | 10 - 11 | 1075 | 6834768 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |