Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008434.2 | NP_032460.2 | 12 - 13 | 1693 | 69 | |
GenBank mRNA | AK136389.1 | - | 5 - 6 | 563 | 69 | |
AK154244.1 | - | 12 - 13 | 1667 | 69 | ||
AY331142.1 | - | 12 - 13 | 1608 | 69 | ||
BC045142.1 | - | 12 - 13 | 1693 | 6314642 | 69 | |
BC055304.1 | - | 12 - 13 | 1648 | 6397978 | 69 | |
U70068.1 | - | 12 - 13 | 1499 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |