Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159527.1 | NP_001152999.1 | 20 - 21 | 2574 | 81 | |
NM_172470.3 | NP_766058.3 | 21 - 22 | 2607 | 81 | ||
GenBank mRNA | AK029896.1 | - | 20 - 21 | 2529 | 81 | |
AK173151.1 | - | 21 - 22 | 2549 | 81 | ||
BC056925.1 | - | 21 - 22 | 2716 | 5704780 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |