Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159527.1 | NP_001152999.1 | 3 - 4 | 363 | 110 | |
NM_172470.3 | NP_766058.3 | 3 - 4 | 363 | 110 | ||
GenBank mRNA | AK015490.1 | - | 3 - 4 | 328 | 110 | |
AK029896.1 | - | 3 - 4 | 318 | 110 | ||
AK031061.1 | - | 3 - 4 | 354 | 110 | ||
AK053724.1 | - | 3 - 4 | 318 | 110 | ||
AK083971.1 | - | 3 - 4 | 308 | 110 | ||
AK173151.1 | - | 3 - 4 | 305 | 110 | ||
BC056925.1 | - | 3 - 4 | 472 | 5704780 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |