Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008792.4 | NP_032818.1 | 7 - 8 | 1061 | 81 | |
GenBank mRNA | AF483512.1 | - | 7 - 8 | 703 | 81 | |
AF483513.1 | - | 7 - 8 | 703 | 81 | ||
AK133620.1 | - | 7 - 8 | 1062 | 81 | ||
AK158493.1 | - | 7 - 8 | 1009 | 81 | ||
AK158598.1 | - | 7 - 8 | 1123 | 81 | ||
AK161585.1 | - | 7 - 8 | 1002 | 81 | ||
BC052013.1 | - | 7 - 8 | 978 | 5707923 | 81 | |
BC057348.1 | - | 7 - 8 | 1016 | 6827695 | 81 | |
M55669.1 | - | 7 - 8 | 787 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |