Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130412.1 | NP_001123884.1 | 7 - 8 | 1054 | 88 | |
NM_015763.4 | NP_056578.2 | 7 - 8 | 1009 | 88 | ||
NM_172950.3 | NP_766538.2 | 6 - 7 | 910 | 88 | ||
GenBank mRNA | AB093214.1 | - | 2 - 3 | 229 | 88 | |
AF412811.1 | - | 6 - 7 | 947 | 88 | ||
AK014526.1 | - | 6 - 7 | 970 | 88 | ||
AK019539.1 | - | 7 - 8 | 1147 | 88 | ||
AK030921.1 | - | 6 - 7 | 913 | 88 | ||
AK141489.1 | - | 7 - 8 | 971 | 88 | ||
AK154837.1 | - | 7 - 8 | 1051 | 88 | ||
BC042462.1 | - | 6 - 7 | 963 | 4211202 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |