Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130412.1 | NP_001123884.1 | 10 - 11 | 1644 | 63 | |
NM_015763.4 | NP_056578.2 | 10 - 11 | 1599 | 63 | ||
NM_172950.3 | NP_766538.2 | 9 - 10 | 1500 | 63 | ||
GenBank mRNA | AB093214.1 | - | 5 - 6 | 819 | 63 | |
AF180471.1 | - | 9 - 10 | 1479 | 63 | ||
AF412811.1 | - | 9 - 10 | 1537 | 63 | ||
AK014526.1 | - | 9 - 10 | 1560 | 63 | ||
AK019539.1 | - | 10 - 11 | 1737 | 63 | ||
AK030921.1 | - | 9 - 10 | 1503 | 63 | ||
AK154837.1 | - | 10 - 11 | 1641 | 63 | ||
BC042462.1 | - | 9 - 10 | 1553 | 4211202 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |