Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134040.1 | NP_598801.1 | 24 - 25 | 2030 | 67 | |
GenBank mRNA | AK028341.1 | - | 24 - 25 | 2027 | 67 | |
AK132157.1 | - | 24 - 25 | 2070 | 67 | ||
AK160982.1 | - | 24 - 25 | 2031 | 67 | ||
AK166146.1 | - | 24 - 25 | 2013 | 67 | ||
BC008570.1 | - | 24 - 25 | 2008 | 3587491 | 67 | |
BC010624.1 | - | 24 - 25 | 2030 | 3153029 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |