Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018828.2 | NP_061298.1 | 15 - 16 | 3076 | 89 | |
GenBank mRNA | AK013152.1 | - | 1 - 2 | 139 | 89 | |
AK084018.1 | - | 15 - 16 | 2868 | 89 | ||
AK129263.1 | - | 14 - 15 | 3947 | 89 | ||
U40750.2 | - | 15 - 16 | 3076 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |