Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200000690.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000690.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000690.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160220.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160220.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160221.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_001160222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000694.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000682.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000682.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000682.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000681.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000681.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000681.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NM_001160219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NM_001160219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NM_001160220.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NM_001160220.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000678.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000675.mm.1 |
NM_001160221.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000675.mm.1 |
NM_001160221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000675.mm.1 |
NM_001160222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000675.mm.1 |
NM_001160222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000675.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160220.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160220.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160221.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_001160222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000689.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NM_001160219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NM_001160219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NM_001160220.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NM_001160220.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000679.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000683.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000683.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000683.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000691.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000691.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000691.mm.1 |
TC1200000068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160219.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160220.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160220.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160221.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_001160222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_030723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NM_030723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NR_027670.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
NR_027670.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200000692.mm.1 |
TC1200000068.mm.1 |
MTA TC |