Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141946.1 | NP_001135418.1 | 3 - 4 | 1744 | 128 | |
NM_001286981.1 | NP_001273910.1 | 4 - 5 | 1776 | 128 | ||
NM_021527.2 | NP_067502.2 | 3 - 4 | 1980 | 128 | ||
GenBank mRNA | AF254074.1 | - | 3 - 4 | 1752 | 128 | |
AK004987.1 | - | 4 - 5 | 1533 | 128 | ||
AK032528.1 | - | 3 - 4 | 1742 | 128 | ||
AK032554.1 | - | 3 - 4 | 1742 | 128 | ||
BC080765.1 | - | 3 - 4 | 1720 | 128 | ||
BC117835.1 | - | 1 - 2 | 1307 | 128 | ||
BC117836.1 | - | 1 - 2 | 1308 | 40093115 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |