Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031811.2 | NP_001026981.2 | 14 - 15 | 2637 | 57 | |
GenBank mRNA | AK048629.1 | - | 14 - 15 | 2526 | 57 | |
BC029690.1 | - | 10 - 11 | 1864 | 57 | ||
BC130020.1 | - | 3 - 4 | 775 | 57 | ||
BC152323.1 | - | 3 - 4 | 775 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |