Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161111.1 | NP_001154583.1 | 2 - 3 | 384 | 84 | |
NM_172574.2 | NP_766162.2 | 2 - 3 | 384 | 84 | ||
GenBank mRNA | AK053171.1 | - | 2 - 3 | 333 | 84 | |
AK089230.1 | - | 2 - 3 | 293 | 84 | ||
AK138680.1 | - | 2 - 3 | 383 | 84 | ||
AK150094.1 | - | 2 - 3 | 296 | 84 | ||
BC025220.1 | - | 2 - 3 | 322 | 5025905 | 84 | |
BY172413.1 | - | 2 - 3 | 333 | 84 | ||
BY208585.1 | - | 2 - 3 | 333 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |