Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027959.3 | NP_082235.1 | 4 - 5 | 383 | 97 | |
GenBank mRNA | AK005989.1 | - | 4 - 5 | 389 | 97 | |
AK133792.1 | - | 4 - 5 | 390 | 97 | ||
AK151008.1 | - | 4 - 5 | 364 | 97 | ||
AK152960.1 | - | 4 - 5 | 396 | 97 | ||
AK165877.1 | - | 4 - 5 | 362 | 97 | ||
AK166111.1 | - | 4 - 5 | 390 | 97 | ||
AK167386.1 | - | 4 - 5 | 401 | 97 | ||
AK168136.1 | - | 4 - 5 | 390 | 97 | ||
AK168279.1 | - | 4 - 5 | 390 | 97 | ||
BC006865.1 | - | 4 - 5 | 372 | 2645183 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |