Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081090.1 | NP_001074559.1 | 12 - 13 | 2223 | 86 | |
GenBank mRNA | AK132230.1 | - | 11 - 12 | 2145 | 86 | |
BC032932.1 | - | 2 - 3 | 131 | 86 | ||
BC150893.1 | - | 12 - 13 | 2189 | 9087804 | 86 | |
BC157979.1 | - | 12 - 13 | 2189 | 9007299 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |