Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009776.3 | NP_033906.2 | 3 - 4 | 659 | 65 | |
GenBank mRNA | AF010254.1 | - | 3 - 4 | 634 | 65 | |
AK144292.1 | - | 2 - 3 | 620 | 65 | ||
AK165744.1 | - | 2 - 3 | 607 | 65 | ||
BC002026.1 | - | 2 - 3 | 590 | 3485810 | 65 | |
Y10386.1 | - | 3 - 4 | 616 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |