Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024856.2 | NP_001020027.1 | 11 - 12 | 1480 | 158 | |
NM_001024857.2 | NP_001020028.1 | 10 - 11 | 1406 | 158 | ||
NM_080788.3 | NP_542966.2 | 11 - 12 | 1234 | 158 | ||
GenBank mRNA | AK046978.1 | - | 11 - 12 | 1453 | 158 | |
AK053820.1 | - | 11 - 12 | 1469 | 158 | ||
AK077011.1 | - | 11 - 12 | 1255 | 158 | ||
AK137024.1 | - | 10 - 11 | 1395 | 158 | ||
AK148269.1 | - | 10 - 11 | 1410 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |