Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 16 - 17 | 2050 | 88 | |
GenBank mRNA | AF211864.1 | - | 16 - 17 | 2050 | 88 | |
AK131133.1 | - | 16 - 17 | 2056 | 88 | ||
AK155816.1 | - | 16 - 17 | 2072 | 88 | ||
AK170432.1 | - | 16 - 17 | 2068 | 88 | ||
BC042840.1 | - | 16 - 17 | 2049 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |