Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sestd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175465.6 | NP_780674.1 | 10 - 11 | 1030 | 70 | |
GenBank mRNA | AK047939.1 | - | 10 - 11 | 1028 | 70 | |
BC013245.1 | - | 11 - 12 | 1111 | 70 | ||
BC044853.1 | - | 10 - 11 | 1030 | 3978465 | 70 | |
CO425092.1 | - | 4 - 5 | 463 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |