Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285831.1 | NP_001272760.1 | 5 - 6 | 711 | 87 | |
NM_027379.3 | NP_081655.2 | 5 - 6 | 711 | 87 | ||
GenBank mRNA | AK005531.1 | - | 5 - 6 | 685 | 87 | |
AK011187.1 | - | 5 - 6 | 686 | 87 | ||
AK014486.1 | - | 5 - 6 | 684 | 87 | ||
AK030067.1 | - | 5 - 6 | 642 | 87 | ||
AK033674.1 | - | 5 - 6 | 654 | 87 | ||
BC007178.1 | - | 5 - 6 | 659 | 3495305 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |