Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdsub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159636.1 | NP_001153108.1 | 6 - 7 | 1031 | 69 | |
NM_028118.2 | NP_082394.1 | 6 - 7 | 1031 | 69 | ||
GenBank mRNA | AK011391.1 | - | 6 - 7 | 967 | 69 | |
BC050792.1 | - | 6 - 7 | 972 | 69 | ||
BC061948.1 | - | 6 - 7 | 1009 | 6312489 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |