Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029832.2 | NP_084108.1 | 3 - 4 | 496 | 84 | |
GenBank mRNA | AK011170.1 | - | 3 - 4 | 427 | 84 | |
BC051541.1 | - | 1 - 2 | 85 | 84 | ||
BC115616.2 | - | 3 - 4 | 496 | 40102754 | 84 | |
BC115617.1 | - | 3 - 4 | 496 | 40102757 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |