Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025576.2 | NP_079852.1 | 1 - 2 | 444 | 56 | |
GenBank mRNA | AK012674.1 | - | 1 - 2 | 434 | 56 | |
AK014691.1 | - | 1 - 2 | 438 | 56 | ||
AK151972.1 | - | 1 - 2 | 445 | 56 | ||
AK168416.1 | - | 252 | 56 | |||
BC026750.1 | - | 1 - 2 | 423 | 5135841 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |