Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010761.2 | NP_034891.2 | 2 - 3 | 265 | 74 | |
GenBank mRNA | AK144883.1 | - | 2 - 3 | 242 | 74 | |
AK154285.1 | - | 2 - 3 | 219 | 74 | ||
AK160205.1 | - | 2 - 3 | 231 | 74 | ||
AK163150.1 | - | 2 - 3 | 178 | 74 | ||
AK167345.1 | - | 2 - 3 | 230 | 74 | ||
BC001984.1 | - | 2 - 3 | 201 | 3489973 | 74 | |
BC005802.1 | - | 1 - 2 | 274 | 3707919 | 74 | |
CO810054.1 | - | 2 - 3 | 272 | 74 | ||
U50734.1 | - | 2 - 3 | 183 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |