Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020568.3 | NP_065593.2 | 2 - 3 | 178 | 106 | |
GenBank mRNA | AF064748.1 | - | 2 - 3 | 189 | 106 | |
AK173286.1 | - | 2 - 3 | 98 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |