Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290656.1 | NP_001277585.1 | 7 - 8 | 4475 | 69 | |
NM_029924.2 | NP_084200.1 | 14 - 15 | 6229 | 69 | ||
GenBank mRNA | AK173176.1 | - | 7 - 8 | 5096 | 69 | |
BC138375.1 | - | 7 - 8 | 4380 | 8861395 | 69 | |
BC138378.1 | - | 7 - 8 | 4380 | 8861398 | 69 | |
BU610422.1 | - | 1 - 2 | 324 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |