Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027921.1 | NP_082197.1 | 3 - 4 | 646 | 82 | |
GenBank mRNA | AK049846.1 | - | 3 - 4 | 698 | 82 | |
BC023456.1 | - | 3 - 4 | 646 | 5041164 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |